Learn More
CTCFL Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant CTCFL.
Brand: Abnova PAB20061.100uL
Description
CCCTC-binding factor (CTCF), an 11-zinc-finger factor involved in gene regulation, utilizes different zinc fingers to bind varying DNA target sites. CTCF forms methylation-sensitive insulators that regulate X-chromosome inactivation. This gene is a paralog of CTCF and appears to be expressed primarily in the cytoplasm of spermatocytes, unlike CTCF which is expressed primarily in the nucleus of somatic cells. CTCF and the protein encoded by this gene are normally expressed in a mutually exclusive pattern that correlates with resetting of methylation marks during male germ cell differentiation. [provided by RefSeq
Sequence: DGVCREKDHRSPSELEAQRTSGAFQDSVLEEEVELVLAPSEESEKYILTLQTVHFTSEAVELQDMSLLSIQQQEGVQVVVQQPGPGLLWLEEGPRQSLQQCVAISIQQELYSPQEMEVLQFHALEENVMVASEDSKLAVSSpecifications
| CTCFL | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant CTCFL. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q8NI51 | |
| CTCFL | |
| Recombinant protein corresponding to amino acids of human CTCFL. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| CTCFL | |
| BORIS/CTCF-T/MGC163358/dJ579F20.2 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 140690 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only