missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CUL3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CUL3.
Brand: Abnova PAB10037.100uL
This item is not returnable.
View return policy
Description
CUL3 is a component of a ubiquitin E3 ligase that is essential for mitotic division (Sumara et al., 2007 [PubMed 17543862]).[supplied by OMIM
Specifications
| CUL3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of CUL3. | |
| In antiserum (0.01% sodium azide) | |
| NP_003581 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Liquid |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| ELISA (1:2000-1:10000) Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
| CUL3 | |
| CUL3 | |
| A synthetic peptide (conjugated with KLH) corresponding to amino acids 1-12 of human CUL3. | |
| RUO | |
| 8452 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction