Learn More
CXCL10 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CXCL10.
Brand: Abnova PAB19527.100ug
Description
This gene encodes a chemokine of the CXC subfamily and ligand for the receptor CXCR3. Binding of this protein to CXCR3 results in pleiotropic effects, including stimulation of monocytes, natural killer and T-cell migration, and modulation of adhesion molecule expression. [provided by RefSeq
Specifications
| CXCL10 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of CXCL10. | |
| Lyophilized from 0.9mg NaCl, 0.2mg Na2HPO4 (5mg BSA, 0.05mg sodium azide, 0.05mg Thimerosal) | |
| C7/IFI10/INP10/IP-10/SCYB10/crg-2/gIP-10/mob-1 | |
| Rabbit | |
| Immunoaffinity purification | |
| RUO | |
| 3627 | |
| Human | |
| Lyophilized |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| CXCL10 | |
| CXCL10 | |
| A synthetic peptide corresponding to amino acids at C-terminus of human CXCL10. | |
| 100 μg | |
| Primary | |
| It identical to the related mouse and rat sequences. | |
| Store at -20°C on dry atmosphere. After reconstitution with 200μL of deionized water and concentration will be 500μ/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only