missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-CXCR3, Polyclonal, Abnova™
Brand: Abnova PAB26088.50ug
This item is not returnable.
View return policy
Specifications
| CXCR3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of CXCR3. | |
| In PBS (0.09% sodium azide) | |
| P49682 | |
| CXCR3 | |
| A synthetic peptide corresponding to 17 amino acids at C-terminus of human CXCR3. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins, except SRRM2 (59%). | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (16 ug/mL) The optimal working dilution should be determined by the end user. | |
| CXCR3 | |
| CD182/CD183/CKR-L2/CMKAR3/GPR9/IP10-R/Mig-R/MigR | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 2833 | |
| Human, Monkey | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction