Learn More
CXCR4 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CXCR4.
Brand: Abnova PAB0361.100ug
Description
This gene encodes a CXC chemokine receptor specific for stromal cell-derived factor-1. The protein has 7 transmembrane regions and is located on the cell surface. It acts with the CD4 protein to support HIV entry into cells and is also highly expressed in breast cancer cells. Mutations in this gene have been associated with WHIM (warts, hypogammaglobulinemia, infections, and myelokathexis) syndrome. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq
Specifications
| CXCR4 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of CXCR4. | |
| In PBS (0.05% BSA, 0.05% sodium azide) | |
| CD184/D2S201E/FB22/HM89/HSY3RR/LAP3/LCR1/LESTR/NPY3R/NPYR/NPYRL/NPYY3R/WHIM | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| CXCR4 | |
| CXCR4 | |
| A synthetic peptide corresponding to amino acids 328-338 of human CXCR4. | |
| RUO | |
| 7852 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only