Learn More
CYLD, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CYLD.
Brand: Abnova PAB12454.100uL
Description
This gene is encodes a cytoplasmic protein with three cytoskeletal-associated protein-glycine-conserved (CAP-GLY) domains that functions as a deubiquitinating enzyme. Mutations in this gene have been associated with cylindromatosis, multiple familial trichoepithelioma, and Brooke-Spiegler syndrome. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq
Specifications
| CYLD | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of CYLD. | |
| In Tris-citrate/phosphate buffer, pH 7.0-8.0 (0.09% sodium azide) | |
| Q9NQC7 | |
| CYLD | |
| A synthetic peptide corresponding to amino acids 500-600 of human CYLD. | |
| RUO | |
| 1540 | |
| Store at 4°C. Do not freeze. |
| Western Blot | |
| Unconjugated | |
| Western Blot (2 mg/mL) The optimal working dilution should be determined by the end user. | |
| CYLD | |
| CDMT/CYLD1/CYLDI/EAC/FLJ20180/FLJ31664/FLJ78684/HSPC057/KIAA0849/MFT/MFT1/SBS/TEM/USPL2 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Bovine, Human, Mouse, Primate | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only