missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYTSB Rabbit anti-Human, Biotin, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CYTSB.
Brand: Abnova PAB14985.100uL
This item is not returnable.
View return policy
Specifications
| CYTSB | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of CYTSB. | |
| In PBS | |
| FLJ36955/HCMOGT-1/NSP/SPECC1 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| This antibody reacts with human protein. There is 83% homology of the immunogen with the mouse and rat NSP5a3a proteins. This is specific to NSP5a3a. This antibody is useful for Western blot analysis, where a band is seen ∼80 KDa. A second band may be seen ∼60 KDa. The specificity of this band is not known. No other applications have been tested. Optimal working dilutions should be determined by the investigator. | |
| Store at 4°C. Do not freeze. |
| Western Blot | |
| Biotin | |
| Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. | |
| CYTSB | |
| CYTSB | |
| A synthetic peptide corresponding to amino acids 1-100 of human CYTSB. | |
| RUO | |
| 92521 | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction