Learn More
DAB1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of DAB1.
Brand:  Abnova PAB18465.100ug
Product Code. 16172970
  • € 406.00 / 100µg
Description
The laminar organization of multiple neuronal types in the cerebral cortex is required for normal cognitive function. In mice, the disabled-1 gene plays a central role in brain development, directing the migration of cortical neurons past previously formed neurons to reach their proper layer. This gene is similar to disabled-1, and the protein encoded by this gene is thought to be a signal transducer that interacts with protein kinase pathways to regulate neuronal positioning in the developing brain. Alternatively spliced transcript variants of this gene have been reported, but their full length nature has not been determined. [provided by RefSeq
Specifications
Specifications
DAB1
Polyclonal
Unconjugated
Western Blot (1:500-1:1000) Immunofluorescence (1:500-1:1000) ELISA (1:40000) The optimal working dilution should be determined by the end user.
DAB1
DAB1
A synthetic peptide corresponding to residues surrounding Y232 of human DAB1.
100 μg
Primary
This antibody is specific to DAB1.
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
ELISA, Immunofluorescence, Western Blot
1 mg/mL
Rabbit polyclonal antibody raised against synthetic peptide of DAB1.
In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide)
O75553
Rabbit
Affinity purification
RUO
1600
Human, Mouse, Rat