Learn More
DDX58 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of DDX58.
Brand: Abnova PAB3686.400uL
Description
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases which are implicated in a number of cellular processes involving RNA binding and alteration of RNA secondary structure. This gene encodes a protein containing RNA helicase-DEAD box protein motifs and a caspase recruitment domain (CARD). It is involved in viral double-stranded (ds) RNA recognition and the regulation of immune response. [provided by RefSeq
Specifications
| DDX58 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of DDX58. | |
| In PBS (0.09% sodium azide) | |
| NP_055129;O95786 | |
| DDX58 | |
| A synthetic peptide (conjugated with KLH) corresponding to internal region of human DDX58. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| DDX58 | |
| DKFZp434J1111/DKFZp686N19181/FLJ13599/RIG-I | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| 23586 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only