Learn More
DGAT1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of DGAT1.
Brand:  Abnova PAB12449.100uL
Product Code. 16099836
  • € 624.00 / 100µL
Description
The enzyme encoded by this gene utilizes diacylglycerol and fatty acyl CoA as substrates in order to catalyze the final stage of triacylglycerol synthesis. It is also involved in cellular as well as physiological metabolic processes. [provided by RefSeq
Specifications
Specifications
DGAT1
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of DGAT1.
In Tris-glycine, 150mM NaCl (0.05% sodium azide)
O75907
DGAT1
A synthetic peptide corresponding to amino acids 200-300 of human DGAT1.
RUO
8694
Store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Western Blot
Unconjugated
Western Blot (2 ug/mL) The optimal working dilution should be determined by the end user.
DGAT1
ARGP1/DGAT
Rabbit
100 μL
Primary
Bovine, Fish, Goat, Human, Mouse, Primate, Rat, Sheep