missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Disc1 Rabbit anti-Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Disc1.
Brand: Abnova PAB5882.100ug
This item is not returnable.
View return policy
Specifications
| Disc1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of Disc1. | |
| In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide) | |
| Disc1 | |
| A synthetic peptide corresponding to C-terminus of mouse Disc1. | |
| RUO | |
| 244667 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Disc1 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction