Learn More
DLGAP2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of DLGAP2.
Brand: Abnova PAB13041.100ug
Description
The product of this gene is one of the membrane-associated guanylate kinases localized at postsynaptic density in neuronal cells. These kinases are a family of signaling molecules expressed at various submembrane domains and contain the PDZ, SH3 and the guanylate kinase domains. This protein may play a role in the molecular organization of synapses and in neuronal cell signaling. Alternatively spliced transcript variants encoding different isoforms have been identified, but their full-length nature is not known. [provided by RefSeq
Specifications
| DLGAP2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of DLGAP2. | |
| In PBS (0.02% sodium azide) | |
| Q9P1A6 | |
| DLGAP2 | |
| A synthetic peptide corresponding to internal region 16 amino acids of human DLGAP2. | |
| RUO | |
| 9228 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| DLGAP2 | |
| DAP2/SAPAP2 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only