missing translation for 'onlineSavingsMsg'
Learn More

DNAAF2 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Product Code. 16173260
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16173260 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16173260 Supplier Abnova Supplier No. PAB20137.100uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against recombinant DNAAF2.

The KTU gene encodes a protein conserved in animals and ciliated unicellular organisms involved in preassembly of dynein arm complexes in species from algae to humans (Omran et al., 2008 [PubMed 19052621]).[supplied by OMIM

Sequence: LQGKEERVNEESHLTEKEYIEHCNTPTTDSDSSIAVKALQIDSFGLVTCFQQESLDVSQMILGKSQQPESKMQSEFIKEKSATCSNEEKGNLNESVITEEKETDGDHLSSLLNKTTVHNIPGFDSIKETNMQDGSVQVIK

Specifications

Antigen DNAAF2
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant DNAAF2.
Dilution Immunohistochemistry (1:200-1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene DNAAF2
Gene Accession No. Q9NVR5
Gene Alias C14orf104/CILD10/KTU/PF13
Gene Symbols DNAAF2
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human DNAAF2.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55172
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.