missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dnahc6, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against partial recombinant Dnahc6.
Brand: Abnova PAB15850.50ug
This item is not returnable.
View return policy
Specifications
| Dnahc6 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against partial recombinant Dnahc6. | |
| In PBS (50% glycerol, 0.02% sodium azide) | |
| AK220414 | |
| Dnahc6 | |
| Recombinant GST fusion protein corresponding to 143 amino acids of mouse Dnahc6. | |
| RUO | |
| 330355 | |
| Mouse | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| Dnahc6 | |
| 9830168K20/A730004I20Rik/KIAA1697/mKIAA1697/mdhc6 | |
| Rabbit | |
| 50 μg | |
| Primary | |
| Specific to recombinant protein GX2241. This antibody detects mDNAH6 protein. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction