missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dock4, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against partial recombinant Dock4.
Brand: Abnova PAB15726.50ug
This item is not returnable.
View return policy
Description
Sequence: CLSPRDRPCSAIYPTPVEPSQRMLFNHIGDGALPRSDPNLSAPEKAVNPTPSSWSLDSGKEAKNMSDSGKLISPPVPPRPTQTASPARHTTSVSPSPAGRSPLKGSVQSFTPSPVEYNSPGLSSNSPVLSGSYSSGISSLSRCSTSETSGFENQANEQSVPVPVPVPVPVPVPSFSGSEEPVRKESKTPPPYSVYERTLRRPVPLPHSLSIPVTSEPPALPPKPLAARSSHLENGTRRTEPGPRPRPLPRKVSQSpecifications
| Dock4 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against partial recombinant Dock4. | |
| In PBS (50% glycerol, 0.02% sodium azide) | |
| AK122353 | |
| Dock4 | |
| Recombinant GST fusion protein corresponding to 254 mouse Dock4. | |
| RUO | |
| 238130 | |
| Mouse | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| Dock4 | |
| 5330406C03/6330411N01Rik/AF263288/C030023J22/mKIAA0716 | |
| Rabbit | |
| 50 μg | |
| Primary | |
| Specific to recombinant protein GX0390. This antibody detects mDOCK4 protein. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction