Learn More
DYRK3 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of DYRK3.
Brand: Abnova PAB2930.400uL
Description
This gene product belongs to the DYRK family of dual-specificity protein kinases that catalyze autophosphorylation on serine/threonine and tyrosine residues. The members of this family share structural similarity, however, differ in their substrate specificity, suggesting their involvement in different cellular functions. The encoded protein has been shown to autophosphorylate on tyrosine residue and catalyze phosphorylation of histones H3 and H2B in vitro. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq
Specifications
| DYRK3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of DYRK3. | |
| In PBS (0.09% sodium azide) | |
| O43781 | |
| DYRK3 | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human DYRK3. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| DYRK3 | |
| DYRK5/RED/REDK/hYAK3-2 | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 8444 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only