missing translation for 'onlineSavingsMsg'
Learn More

EGF Rabbit anti-Human, Polyclonal Antibody, Abnova™

Product Code. 16133030
Change view
Click to view available options
Quantity:
150 μg
Unit Size:
150µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16133030 150 μg 150µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16133030 Supplier Abnova Supplier No. PAB19853.150ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against recombinant EGF.

Epidermal growth factor has a profound effect on the differentiation of specific cells in vivo and is a potent mitogenic factor for a variety of cultured cells of both ectodermal and mesodermal origin. The EGF precursor is believed to exist as a membrane-bound molecule which is proteolytically cleaved to generate the 53-amino acid peptide hormone that stimulates cells to divide. [provided by RefSeq

Specifications

Antigen EGF
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant EGF.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (0.02% sodium azide, 50% glycerol (v/v))
Gene EGF
Gene Alias HOMG4/URG
Gene Symbols EGF
Host Species Rabbit
Immunogen Recombinant protein corresponding to human EGF.
Purification Method Affinity purification
Quantity 150 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1950
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.