missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF2AK2 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of EIF2AK2.
Brand: Abnova PAB25180.100uL
This item is not returnable.
View return policy
Specifications
| EIF2AK2 | |
| Polyclonal | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| EIF2AK2 | |
| EIF2AK2 | |
| A synthetic peptide corresponding to EIF2AK2. | |
| 100 μL | |
| Primary | |
| EIF2AK2 polyclonal antibody detects endogenous levels of EIF2AK2 protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed) | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of EIF2AK2. | |
| In PBS, pH 7.2 (0.05% sodium azide) | |
| EIF2AK1/MGC126524/PKR/PRKR | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 5610 | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction