missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPHA2/EPHA3/EPHA4 (phospho Y588/Y596), Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Specifications
Specifications
| Antigen | EPHA2/EPHA3/EPHA4 |
| Applications | Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Rabbit polyclonal antibody raised against synthetic phosphopeptide of human EPHA2/EPHA3/EPHA4. |
| Dilution | Immunofluorescence (1:100∼1:200) Western Blot (1:500∼1:1000) The optimal working dilution should be determined by the end user. |
| Formulation | In PBS (without mg2+ and Ca2+), 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) |
| Gene Accession No. | P29317;P29320;P54764 |
| Gene Alias | ECK |
| Gene Symbols | EPHA2 |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?