Learn More
EPM2A Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of EPM2A.
Brand: Abnova PAB3106.400uL
Description
This gene encodes a dual-specificity phosphatase that associates with polyribosomes. The encoded protein may be involved in the regulation of glycogen metabolism. Mutations in this gene have been associated with myoclonic epilepsy of Lafora. Alternative splicing results in multiple transcript variants. [provided by RefSeq
Specifications
| EPM2A | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of EPM2A. | |
| In PBS (0.09% sodium azide) | |
| NP_005661;O95278 | |
| EPM2A | |
| A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human EPM2A. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) Immunohistochemistry (1:10-50) The optimal working dilution should be determined by the end user. | |
| EPM2A | |
| EPM2/MELF | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 7957 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only