missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Equine IgG (H+L) Secondary Antibody, Janelia Fluor 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB120-6700JF646
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Equine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| IgG (H+L) | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Horse IgG whole molecule | |
| 0.5 mL | |
| Adaptive Immunity, Immunology | |
| Equine | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Janelia Fluor 646 | |
| Rabbit | |
| Multi-step | |
| RUO | |
| Secondary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction