Learn More
ERAP1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ERAP1.
Marca: Abnova PAB25601.50ug
Descrizione
Aminopeptidases play a role in the metabolism of several peptides that may be involved in blood pressure and the pathogenesis of essential hypertension (MIM 145500). Adipocyte-derived leucine aminopeptidase (ALAP) is a member of the M1 family of zinc metallopeptidases.[supplied by OMIM
Specifica
| ERAP1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of ERAP1. | |
| In PBS (0.09% sodium azide) | |
| A-LAP/ALAP/APPILS/ARTS-1/ARTS1/ERAAP/ERAAP1/KIAA0525/PILS-AP/PILSAP | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 51752 | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| ERAP1 | |
| ERAP1 | |
| A synthetic peptide corresponding to 15 amino acids at C-terminus of human ERAP1. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
For Research Use Only