Learn More
ERK/MAPK (phospho T202/Y204), Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of ERK/MAPK.
Brand: Abnova PAB9666.100uL
This item is not returnable.
View return policy
Specifications
| ERK/MAPK | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of ERK/MAPK. | |
| In 10mM HEPES, 150mM NaCl, pH 7.5 (50% glycerol, 10% BSA) | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| Bovine, Canine, Chicken, Fish, Frog, Human, Mouse, Primate, Rat | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| P63086 | |
| Synthetic phosphopeptide corresponding to residues surrounding T202/Y204 of rat ERK/MAPK. | |
| 100 μL | |
| Primary | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |