Learn More
ERP29 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant ERP29.
Brand: Abnova PAB23419.100uL
Description
This gene encodes a reticuloplasmin, a protein which resides in the lumen of the endoplasmic reticulum (ER). The protein shows sequence similarity to the protein disulfide isomerase family. However, it lacks the thioredoxin motif characteristic of this family, suggesting that this protein does not function as a disulfide isomerase. The protein dimerizes and is thought to play a role in the processing of secretory proteins within the ER. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq
Sequence: SYPVFYLFRDGDFENPVPYTGAVKVGAIQRWLKGQGVYLGMPGCLPVYDALAGEFIRASGVEARQALLKQGQDNLSSVSpecifications
| ERP29 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant ERP29. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| P30040 | |
| ERP29 | |
| Recombinant protein corresponding to amino acids of human ERP29. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| ERP29 | |
| C12orf8/ERp28/ERp31/PDI-DB | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 10961 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only