Learn More
ETS1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ETS1.
Brand: Abnova PAB24790.100uL
Description
ETS transcriptions factors, such as ETS1, regulate numerous genes and are involved in stem cell development, cell senescence and death, and tumorigenesis. The conserved ETS domain within these proteins is a winged helix-turn-helix DNA-binding domain that recognizes the core consensus DNA sequence GGAA/T of target genes (Dwyer et al., 2007 [PubMed 17986575]).[supplied by OMIM
Specifications
| ETS1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of ETS1. | |
| In PBS, pH 7.2 (0.09% sodium azide) | |
| ETS-1/EWSR2/FLJ10768 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 2113 | |
| Human | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
| ETS1 | |
| ETS1 | |
| A synthetic peptide corresponding to ETS1. | |
| 100 μL | |
| Primary | |
| ETS1 polyclonal antibody detects endogenous levels of ETS1 protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only