Learn More
EZR (phospho T353), Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of EZR.
Brand:  Abnova PAB18517.100ug
Product Code. 16063127
  • € 406.00 / 100µg
Description
The cytoplasmic peripheral membrane protein encoded by this gene functions as a protein-tyrosine kinase substrate in microvilli. As a member of the ERM protein family, this protein serves as an intermediate between the plasma membrane and the actin cytoskeleton. This protein plays a key role in cell surface structure adhesion, migration and organization, and it has been implicated in various human cancers. A pseudogene located on chromosome 3 has been identified for this gene. Alternatively spliced variants have also been described for this gene. [provided by RefSeq
Specifications
Specifications
EZR
Polyclonal
Unconjugated
Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:100) The optimal working dilution should be determined by the end user.
EZR
CVIL/CVL/DKFZp762H157/FLJ26216/MGC1584/VIL2
Rabbit
Affinity purification
RUO
7430
Human, Mouse
Liquid
Immunohistochemistry (PFA fixed), Western Blot
1 mg/mL
Rabbit polyclonal antibody raised against synthetic phosphopeptide of EZR.
In PBS, 150mM NaCl, pH 7.4 (0.02% sodium azide, 50% glycerol).
P15311
EZR
Synthetic phosphopeptide corresponding to residues surrounding T353 of human EZR.
100 μg
Primary
This antibody is specific to EZR only when phosphorylated at T353.
Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.