Learn More
F2 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against native F2.
Brand: Abnova PAB5051.150ug
Description
Coagulation factor II is proteolytically cleaved to form thrombin in the first step of the coagulation cascade which ultimately results in the stemming of blood loss. F2 also plays a role in maintaining vascular integrity during development and postnatal life. Mutations in F2 leads to various forms of thrombosis and dysprothrombinemia. [provided by RefSeq
Specifications
| F2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against native F2. | |
| In PBS, pH 7.5 (50% glycerol, 0.01% thimerosal) | |
| PT | |
| Rabbit | |
| 150 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Immunoprecipitation | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| F2 | |
| F2 | |
| Native purified human F2. | |
| RUO | |
| 2147 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only