Learn More
F2RL1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of F2RL1.
Brand: Abnova PAB26247.50ug
Description
Coagulation factor II (thrombin) receptor-like 1 (F2RL1) is a member of the large family of 7-transmembrane-region receptors that couple to guanosine-nucleotide-binding proteins. F2RL1 is also a member of the protease-activated receptor family. It is activated by trypsin, but not by thrombin. It is activated by proteolytic cleavage of its extracellular amino terminus. The new amino terminus functions as a tethered ligand and activates the receptor. The F2RL1 gene contains two exons and is widely expressed in human tissues. The predicted protein sequence is 83% identical to the mouse receptor sequence. [provided by RefSeq
Specifications
| F2RL1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of F2RL1. | |
| In PBS (0.09% sodium azide) | |
| P55085 | |
| F2RL1 | |
| A synthetic peptide corresponding to 18 amino acid at extracellular domain of human F2RL1. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins, except NMBR (39%). | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) The optimal working dilution should be determined by the end user. | |
| F2RL1 | |
| GPR11/PAR2 | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 2150 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only