missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM102B, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant FAM102B.
Brand: Abnova PAB22155.100uL
This item is not returnable.
View return policy
Description
Sequence: SRSSSFSELCHRRNTSVGSTSTGVESILEPCDEIEQKIAEPNLDTADKEDTASEKLSRCPVKQDSVESQLKRVDDTRVDADDIVEKILQSQDSpecifications
| FAM102B | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant FAM102B. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q5T8I3 | |
| FAM102B | |
| Recombinant protein corresponding to amino acids of human FAM102B. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) The optimal working dilution should be determined by the end user. | |
| FAM102B | |
| DKFZp686N01110/DKFZp779B126 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 284611 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction