Learn More
FANCD2 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against partial recombinant FANCD2.
Brand: Abnova PAB12428.50uL
Description
The Fanconi anemia complementation group (FANC) currently includes FANCA, FANCB, FANCC, FANCD1 (also called BRCA2), FANCD2, FANCE, FANCF, FANCG, FANCI, FANCJ (also called BRIP1), FANCL, FANCM and FANCN (also called PALB2). The previously defined group FANCH is the same as FANCA. Fanconi anemia is a genetically heterogeneous recessive disorder characterized by cytogenetic instability, hypersensitivity to DNA crosslinking agents, increased chromosomal breakage, and defective DNA repair. The members of the Fanconi anemia complementation group do not share sequence similarity; they are related by their assembly into a common nuclear protein complex. This gene encodes the protein for complementation group D2. This protein is monoubiquinated in response to DNA damage, resulting in its localization to nuclear foci with other proteins (BRCA1 AND BRCA2) involved in homology-directed DNA repair. Alternative splicing results in two transcript variants encoding different isoforms. [provided by RefSeq
Specifications
| FANCD2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against partial recombinant FANCD2. | |
| In Tris-glycine, 150mM NaCl (0.05% sodium azide) | |
| Q9BXW9 | |
| FANCD2 | |
| Recombinant protein corresponding to N-terminus of human FANCD2. | |
| RUO | |
| 2177 | |
| Human, Mouse | |
| Liquid |
| Immunofluorescence, Immunohistochemistry, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Immunofluorescence (1:200-1:500) Western Blot (1:10000-1:20000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2.5-5 ug/mL) The optimal working dilution should be determined by the end user. | |
| FANCD2 | |
| DKFZp762A223/FA-D2/FA4/FACD/FAD/FAD2/FANCD/FLJ23826 | |
| Rabbit | |
| 50 μL | |
| Primary | |
| This antibody is specific to FANCD2. | |
| Store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only