missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fap, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Fap.
Brand: Abnova PAB10209.100ug
This item is not returnable.
View return policy
Specifications
| Fap | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of Fap. | |
| In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| NP_032012;P97321 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody is known to react with mouse FAP protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:100000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. | |
| Fap | |
| Fap | |
| A synthetic peptide corresponding to internal region of mouse Fap. | |
| RUO | |
| 14089 | |
| Human, Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction