missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fbxo28, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against partial recombinant Fbxo28.
Brand: Abnova PAB15817.50ug
This item is not returnable.
View return policy
Specifications
| Fbxo28 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against partial recombinant Fbxo28. | |
| In PBS (50% glycerol, 0.02% sodium azide) | |
| AK122295 | |
| Fbxo28 | |
| Recombinant GST fusion protein corresponding to 104 amino acids of mouse Fbxo28. | |
| RUO | |
| 67948 | |
| Mouse | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| Fbxo28 | |
| 4833428J17Rik/5730505P19Rik/D1Ertd578e/Fbx28/KIAA0483/MGC90899/mKIAA0483 | |
| Rabbit | |
| 50 μg | |
| Primary | |
| Specific to recombinant protein GX0462. This antibody detects mFBXO28 protein. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction