missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Feline IgG Fc Secondary Antibody, Allophycocyanin/Cy7, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-72771APCCY7
This item is not returnable.
View return policy
Description
IgG Fc Polyclonal antibody specifically detects IgG Fc in Feline samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Dot Blot
Specifications
| IgG Fc | |
| Polyclonal | |
| PBS | |
| Cat IgG F(c) fragment | |
| 500 μg | |
| Adaptive Immunity, Immunology | |
| Feline | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Dot Blot | |
| APC-Cyanine7 | |
| Rabbit | |
| Multi-step | |
| RUO | |
| Secondary | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction