Learn More
FGR Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant FGR.
Brand: Abnova PAB28540.100uL
Description
This gene is a member of the Src family of protein tyrosine kinases (PTKs). The encoded protein contains N-terminal sites for myristylation and palmitylation, a PTK domain, and SH2 and SH3 domains which are involved in mediating protein-protein interactions with phosphotyrosine-containing and proline-rich motifs, respectively. The protein localizes to plasma membrane ruffles, and functions as a negative regulator of cell migration and adhesion triggered by the beta-2 integrin signal transduction pathway. Infection with Epstein-Barr virus results in the overexpression of this gene. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq
Sequence: VFCKKLEPVATAKEDAGLEGDFRSYGAADHYGPDPTKARPASSFAHIPNYSNFSSQAINPGFLDSGTIRGVSGIGVTLFIALYDYEARTEDDLTFTKGEKFHILNNTEGDWWEARSLSSGKTGCIPSNYVAPVDSIQAEEWYFGKIGSpecifications
| FGR | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant FGR. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| P09769 | |
| FGR | |
| Recombinant protein corresponding to amino acids of human FGR. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1:100-1:250) Immunohistochemistry (1:200-1:500) The optimal working dilution should be determined by the end user. | |
| FGR | |
| FLJ43153/MGC75096/SRC2/c-fgr/c-src2/p55c-fgr/p58c-fgr | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 2268 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only