missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FHL2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of FHL2.
Brand: Abnova PAB9874.100ug
This item is not returnable.
View return policy
Description
LIM proteins contain a highly conserved double zinc finger motif called the LIM domain.[supplied by OMIM
Specifications
| FHL2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of FHL2. | |
| In PBS (0.08% sodium azide) | |
| AAG11/DRAL/SLIM3 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| Western Blot (0.5-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| FHL2 | |
| FHL2 | |
| A synthetic peptide corresponding to human FHL2. | |
| RUO | |
| 2274 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction