missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibrinogen Rabbit anti-Bovine, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against native Fibrinogen.
Brand: Abnova PAB19899.150ug
This item is not returnable.
View return policy
Specifications
| Fibrinogen | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against native Fibrinogen. | |
| In PBS, pH 7.5 (0.02% sodium azide, 50% glycerol (v/v)) | |
| FGA | |
| Native purified fibrinogen from bovine plasma. | |
| 150 μg | |
| Primary | |
| Bovine | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| MGC128011 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 522039/510522/280792 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction