missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FLJ22374 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant FLJ22374.
Brand: Abnova PAB22396.100uL
This item is not returnable.
View return policy
Description
Sequence: VQGCVLQKLLFEGDSKADCAQGLQPSDAHRTRCLVLALADIVWRAGGRERAVVALASRTQQFSPTGKYKADGVLETLTLHSLTCYEDLVTFLQQSIHQFEVGPYGCILLTLSAILSRSTELIRQDFDVPTSSpecifications
| FLJ22374 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant FLJ22374. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q4G0A6 | |
| FLJ22374 | |
| Recombinant protein corresponding to amino acids of human FLJ22374. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| FLJ22374 | |
| MGC44277 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 84182 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction