Learn More
FLOT2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant FLOT2.
Brand: Abnova PAB28427.100uL
Description
Caveolae are small domains on the inner cell membrane involved in vesicular trafficking and signal transduction. This gene encodes a caveolae-associated, integral membrane protein, which is thought to function in neuronal signaling. [provided by RefSeq
Sequence: ECKKEMLDVKFMADTKIADSKRAFELQKSAFSEEVNIKTAEAQLAYELQGAREQQKIRQEEIEIEVVQRKKQIAVEAQEILRTDKELIATVRRPAEAEAHRIQQIAEGEKVKQVLLAQAEAEKIRKSpecifications
| FLOT2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant FLOT2. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| ECS-1/ECS1/ESA/ESA1/M17S1 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 2319 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry(1:2500-1:5000) Western Blot(1:100-1:250) The optimal working dilution should be determined by the end user. | |
| FLOT2 | |
| FLOT2 | |
| Recombinant protein corresponding to amino acids of recombinant FLOT2. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only