missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXO4 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody rasied against synthetic peptide of FOXO4.
Brand: Abnova PAB19264.100ug
This item is not returnable.
View return policy
Specifications
| FOXO4 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (0.5-1 ug/mL) Immunocytochemistry (10 ug/mL) The optimal working dilution should be determined by the end user. | |
| FOXO4 | |
| AFX/AFX1/MGC120490/MLLT7 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 4303 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunocytochemistry, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody rasied against synthetic peptide of FOXO4. | |
| In PBS (0.02% sodium azide) | |
| NP_005929 | |
| FOXO4 | |
| A synthetic peptide corresponding to 15 amino acids near N-terminus of human FOXO4. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction