Learn More
FOXP2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of FOXP2.
Brand: Abnova PAB12684.100ug
Description
This gene encodes an evolutionarily conserved transcription factor expressed in fetal and adult brain. This transcription factor is a member of the forkhead/winged-helix (FOX) family of transcription factors, and contains a FOX DNA-binding domain and a large polyglutamine tract. Members of the FOX family of transcription factors are regulators of embryogenesis. The product of this gene is thought to be required for proper development of speech and language regions of the brain during embryogenesis. Although a point mutation in this gene has been associated with the KE pedigree segregating developmental verbal dyspraxia, no association between mutations in this gene and another speech disorder, autism, has been found. Multiple alternative transcripts encoding different isoforms have been identified. [provided by RefSeq
Specifications
| FOXP2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of FOXP2. | |
| In TBS, pH 7.2 (BSA, 10% Proclin300) | |
| CAGH44/DKFZp686H1726/SPCH1/TNRC10 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody recognizes ∼83 KDa of FOXP2 protein. | |
| Store at 4°C. For long term storage store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| ELISA, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Western Blot (0.5-5 ug/mL) ELISA (0.01-0.1 ug/mL) Immunoprecipitation (2-5 ug/mL) Immunohistochemistry (0.5-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| FOXP2 | |
| FOXP2 | |
| A synthetic peptide corresponding to internal region of human FOXP2. | |
| RUO | |
| 93986 | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only