missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Foxp3 Rabbit anti-Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant Foxp3.
Brand: Abnova PAB12141.100uL
This item is not returnable.
View return policy
Specifications
| Foxp3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant Foxp3. | |
| In PBS (50% glycerol) | |
| Q99JB6 | |
| Foxp3 | |
| Recombinant GST and Flag fusion protein corresponding to amino acids 43-100 of mouse Foxp3. | |
| RUO | |
| 20371 | |
| Store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing. |
| Flow Cytometry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Foxp3 | |
| JM2/scurfin/sf | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction