Learn More
FREQ Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of FREQ.
Brand: Abnova PAB3601.400uL
Description
This gene is a member of the neuronal calcium sensor gene family, which encode calcium-binding proteins expressed predominantly in neurons. The protein encoded by this gene regulates G protein-coupled receptor phosphorylation in a calcium-dependent manner and can substitute for calmodulin. The protein is associated with secretory granules and modulates synaptic transmission and synaptic plasticity. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
| FREQ | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of FREQ. | |
| In PBS (0.09% sodium azide) | |
| P36610 | |
| FREQ | |
| A synthetic peptide (conjugated with KLH) corresponding to internal region of human FREQ. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Flow Cytometry (1:10-50) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-100) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| FREQ | |
| DKFZp761L1223/FLUP/NCS-1/NCS1 | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 23413 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only