Learn More
GAB1 (phospho Y659), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of GAB1.
Brand:  Abnova PAB16972.100ug
Product Code. 16052517
  • € 406.00 / 100µg
Description
The protein encoded by this gene is a member of the IRS1-like multisubstrate docking protein family. It is an important mediator of branching tubulogenesis and plays a central role in cellular growth response, transformation and apoptosis. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
Specifications
GAB1
Polyclonal
Rabbit polyclonal antibody raised against synthetic phosphopeptide of GAB1.
In 20mM PBS, 0.15M NaCl, pH 7.2 (0.01% sodium azide)
Q13480
Rabbit
100 μg
Primary
This antibody detects endogenous levels of GAB1 only when phosphorylated at tyrosine 659.
Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
ELISA, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:100) ELISA (1:4000) The optimal working dilution should be determined by the end user.
GAB1
GAB1
Synthetic phosphopeptide corresponding to residues surrounding Y659 of human GAB1.
RUO
2549
Human, Mouse, Rat