Learn More
GAL3ST1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant GAL3ST1.
Brand: Abnova PAB28396.100uL
Description
Sulfonation, an important step in the metabolism of many drugs, xenobiotics, hormones, and neurotransmitters, is catalyzed by sulfotransferases. The product of this gene is galactosylceramide sulfotransferase which catalyzes the conversion between 3'-phosphoadenylylsulfate + a galactosylceramide to adenosine 3',5'-bisphosphate + galactosylceramide sulfate. Activity of this sulfotransferase is enhanced in renal cell carcinoma. [provided by RefSeq]
Sequence: LLNILFRFGQKHRLKFAFPNGRNDFDYPTFFARSLVQDYRPGACFNIICNHMRFHYDEVRGLVPTNAIFITVLRDPARLFESSFHYFGPVVPLTWKLSAGDKLTEFLQDPDRYYDPNGFNAHYLRNLLFFDLGYDNSLDPSSPQVQEHILSpecifications
| GAL3ST1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant GAL3ST1. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| C9J6M2 | |
| GAL3ST1 | |
| Recombinant protein corresponding to human GAL3ST1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:20-1:50) Immunofluorescence (1-4 ug/ml) The optimal working dilution should be determined by the end user. | |
| GAL3ST1 | |
| CST | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 9514 | |
| Store at 4 °C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only