Learn More
GDF15 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GDF15.
Brand: Abnova PAB19942.100ug
Description
Bone morphogenetic proteins (e.g., BMP5; MIM 112265) are members of the transforming growth factor-beta (see TGFB1; MIM 190180) superfamily and regulate tissue differentiation and maintenance. They are synthesized as precursor molecules that are processed at a dibasic cleavage site to release C-terminal domains containing a characteristic motif of 7 conserved cysteines in the mature protein.[supplied by OMIM
Specifications
| GDF15 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of GDF15. | |
| In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| GDF-15/MIC-1/MIC1/NAG-1/PDF/PLAB/PTGFB | |
| Rabbit | |
| Immunoaffinity purification | |
| RUO | |
| 9518 | |
| Human, Mouse | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:300000) Western Blot (1:2000-1:10000) The optimal working dilution should be determined by the end user. | |
| GDF15 | |
| GDF15 | |
| A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human GDF15. | |
| 100 μg | |
| Primary | |
| This antibody reacts with the C-terminus of endogenous NAG-1 protein from human and mouse tissues. A BLAST analysis suggests reactivity with NAG-1 from chimpanzee and macaque based on a 100% homology. Partial reactivity is expected against rat based on an 86% homology with the immunizing sequence. Cross-reactivity with NAG-1 from other sources has not been determined. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only