Learn More
GDF6 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant GDF6.
Brand: Abnova PAB24393.100uL
Description
This gene encodes a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily of secreted signaling molecules. It is required for normal formation of some bones and joints in the limbs, skull, and axial skeleton. Mutations in this gene result in colobomata, which are congenital abnormalities in ocular development, and in Klippel-Feil syndrome (KFS), which is a congenital disorder of spinal segmentation. [provided by RefSeq
Sequence: IYRTYSIAEKLGINASFFQSSKSANTITSFVDRGLDDLSHTPLRRQKYLFDVSMLSDKEELVGAELRLFRQAPSAPWGPPAGPLHVQLFSpecifications
| GDF6 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant GDF6. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q6KF10 | |
| GDF6 | |
| Recombinant protein corresponding to amino acids of human GDF6. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| GDF6 | |
| BMP13/CDMP2/KFS/KFSL/MGC158100/MGC158101/SGM1 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 392255 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only