Learn More
GLI2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GLI2.
Brand:  Abnova PAB10051.100ug
Product Code. 16019026
  • € 870.00 / 100µg
Description
This gene encodes a protein which belongs to the C2H2-type zinc finger protein subclass of the Gli family. Members of this subclass are characterized as transcription factors which bind DNA through zinc finger motifs. These motifs contain conserved H-C links. Gli family zinc finger proteins are mediators of Sonic hedgehog (Shh) signaling and they are implicated as potent oncogenes in the embryonal carcinoma cell. The protein encoded by this gene localizes to the cytoplasm and activates patched Drosophila homolog (PTCH) gene expression. It is also thought to play a role during embryogenesis. The encoded protein is associated with several phenotypes- Greig cephalopolysyndactyly syndrome, Pallister-Hall syndrome, preaxial polydactyly type IV, postaxial polydactyly types A1 and B. [provided by RefSeq
Specifications
Specifications
GLI2
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of GLI2.
In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
P10070;NP_084655(isoformalpha)
GLI2
A synthetic peptide corresponding to amino acids 46-60 of human GLI2.
RUO
2736
Bovine, Human, Mouse, Rat
Liquid
ELISA, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
ELISA (1:10,000-1:3,0000) Western Blot (1:500-1:2000) Immunohistochemistry (1:500-1:2000) The optimal working dilution should be determined by the end user.
GLI2
HPE9/THP1/THP2
Rabbit
100 μg
Primary
Expect reactivity with all isoforms of this protein.
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.