missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLUL Rabbit anti-Bovine, Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant GLUL.
Brand: Abnova PAB12567.100uL
This item is not returnable.
View return policy
Specifications
| GLUL | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant GLUL. | |
| In antiserum (0.05% sodium azide) | |
| Recombinant protein corresponding to GLUL. | |
| RUO | |
| Bovine, Human, Mouse, Rat | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:25000) Western Blot (1:10000) The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction