Learn More
GMEB2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant GMEB2.
Brand: Abnova PAB20489.100uL
Description
This gene is a member of KDWK gene family. The product of this gene associates with GMEB1 protein, and the complex is essential for parvovirus DNA replication. Study of rat homolog implicates the role of this gene in modulation of transactivation by the glucocorticoid receptor bound to glucocorticoid response elements. This gene appears to use multiple polyadenylation sites. [provided by RefSeq
Sequence: TSVPLGKVVSTLPSTVLGKGSLQAPPASSPASPLLGGYTVLASSGSTYPSTVEIHPDASSLTVLSTAAVQDGSTVFKVVSPLQLLTLPGLGPTLQNVAQASPGSSTIVTVPAGAAPGPEEHTATIESpecifications
| GMEB2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant GMEB2. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q9UKD1 | |
| GMEB2 | |
| Recombinant protein corresponding to amino acids of human GMEB2. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) Western Blot (1:250-1:500) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| GMEB2 | |
| KIAA1269/P79PIF/PIF79 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 26205 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only