missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Goat IgG F(ab')2 Secondary Antibody, Biotin (Pre-adsorbed), Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-73577
This item is not returnable.
View return policy
Description
IgG F(ab')2 Polyclonal antibody specifically detects IgG F(ab')2 in Goat samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| IgG F(ab')2 | |
| Polyclonal | |
| Biotin | |
| immunoglobulin heavy constant gamma 1 (G1m marker) | |
| Goat IgG F(ab')2 fragment | |
| 1 mg | |
| Secondary | |
| Reconstitute with 1.0 ml deionized water (or equivalent). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| LYOPH | |
| Western Blot 1:2000 - 1:10000, ELISA 1:20000 - 1:100000, Immunohistochemistry 1:1000 - 1:5000 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| This antibody was pre-adsorbed against Human Serum Proteins. No reaction was observed against Goat IgG F(c) or Human Serum Proteins. | |
| Goat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction